Gene Transcripts
- Gene Symbol
- SH3BP2
- Entrez Gene ID
- 6452
- Gene Name
- SH3 domain binding protein 2
- Gene Aliases
- 3BP-2, 3BP2, CRBM, CRPM, RES4-23
- Chromosome Location
- Chr. 4: 2793023 - 2841096 on Build GRCh38
- UniGene ID
- Hs.167679
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001122681.1 |
NP_001116153.1
|
7, 8 |
706 |
|
NM_001145855.1 |
NP_001139327.1
|
7, 8 |
849 |
|
NM_001145856.1 |
NP_001139328.1
|
7, 8 |
796 |
|
NM_003023.4 |
NP_003014.3
|
7, 8 |
847 |
|
GenBank mRNA |
AB000462.1 |
- |
7, 8 |
847 |
|
AB000463.1 |
- |
8, 9 |
1026 |
|
AF000936.1 |
- |
6, 7 |
586 |
|
AK124073.1 |
- |
2, 3 |
692 |
|
AK126537.1 |
- |
2, 3 |
1063 |
|
AK225497.1 |
- |
7, 8 |
708 |
|
AK299996.1 |
- |
7, 8 |
765 |
|
AK312286.1 |
- |
7, 8 |
644 |
|
BC022996.1 |
- |
7, 8 |
702 |
5103986 |
U56386.1 |
- |
7, 8 |
794 |
|
View Full Product Details