Gene Transcripts

Gene Symbol
SPCS1
Entrez Gene ID
28972
Gene Name
signal peptidase complex subunit 1
Gene Aliases
HSPC033, SPC1, SPC12, YJR010C-A
Chromosome Location
Chr. 3: 52705841 - 52708182 on Build GRCh38
UniGene ID
Hs.11125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014041.3 NP_054760.3 4 795
GenBank mRNA AF092138.1 - 4 524
AK027426.1 - 4 524
AX775787.1 - 4 504
AX775833.1 - 4 514
BC000884.1 - 4 504 3462307
BI826431.1 - 4 489
BQ423345.1 - 4 531
L38852.1 - 4 514

View Full Product Details