Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 11 | 1448 | |
GenBank mRNA | AF155658.1 | - | 11 | 1396 | |
AF161466.1 | - | 11 | 1407 | ||
AK297826.1 | - | 11 | 1325 | ||
AK297908.1 | - | 606 | |||
AK312503.1 | - | 11 | 1408 | ||
AL050255.1 | - | 11 | 1392 | ||
AL137272.1 | - | 4 | 394 | ||
BC000151.1 | - | 11 | 1386 | 2984824 | |
BC002970.1 | - | 11 | 1386 | 3533883 | |
BC010308.1 | - | 11 | 1398 | 3451576 | |
BC016707.1 | - | 11 | 1413 | 4102148 | |
CR456450.1 | - | 11 | 1363 | ||
CR457175.1 | - | 11 | 1317 |