Gene Transcripts

Gene Symbol
LZTS3
Entrez Gene ID
9762
Gene Name
leucine zipper tumor suppressor family member 3
Gene Aliases
PROSAPIP1
Chromosome Location
Chr. 20: 3162617 - 3173597 on Build GRCh38
UniGene ID
Hs.90232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282533.1 NP_001269462.1 4 1125
XM_005260890.3 XP_005260947.1 4 1069
XM_005260892.1 XP_005260949.1 4 1125
XM_005260893.2 XP_005260950.1 4 1038
XM_011529407.2 XP_011527709.1 4 1069
XM_011529408.2 XP_011527710.1 4 2107
XM_011529409.2 XP_011527711.1 4 2527
GenBank mRNA AB011124.1 - 2 2153
BC038860.1 - 4 1081 5260571

View Full Product Details