Gene Transcripts

Gene Symbol
ORMDL1
Entrez Gene ID
94101
Gene Name
ORMDL sphingolipid biosynthesis regulator 1
Gene Aliases
Chromosome Location
Chr. 2: 189766149 - 189784371 on Build GRCh38
UniGene ID
Hs.700632
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128150.1 NP_001121622.1 4 634
NM_016467.4 NP_057551.1 5 744
XM_005246970.2 XP_005247027.1 4 663
XM_017005347.1 XP_016860836.1 5 783
GenBank mRNA AB064959.1 - 3 1203
AB064960.1 - 4 642
AF151036.1 - 4 582
AF395704.1 - 5 699
AK074756.1 - 4 633
AK075160.1 - 3 635
AK126336.1 - 1 3669
BC005200.1 - 4 601 3929844
CR936635.1 - 3 1311

View Full Product Details