Gene Transcripts

Gene Symbol
PID1
Entrez Gene ID
55022
Gene Name
phosphotyrosine interaction domain containing 1
Gene Aliases
HMFN2073, NYGGF4, P-CLI1, PCLI1
Chromosome Location
Chr. 2: 229023973 - 229271341 on Build GRCh38
UniGene ID
Hs.745038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100818.1 NP_001094288.1 3 2055
NM_017933.4 NP_060403.3 4 2266
XM_011511384.2 XP_011509686.1 3 1938
XM_017004404.1 XP_016859893.1 4 7197
XM_017004405.1 XP_016859894.1 5 2136
XM_017004406.1 XP_016859895.1 3 1794
XM_017004407.1 XP_016859896.1 2 1851
GenBank mRNA AB075874.1 - 4 1921
AK000708.1 - 4 1730
AK096636.1 - 3 2000
AK125359.1 - 4 2190
BC040164.1 - 2 1842 5589172

View Full Product Details