Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001308164.1 | NP_001295093.1 | 16 | 2020 | |
NM_024996.5 | NP_079272.4 | 15 | 1963 | ||
XM_006713795.1 | XP_006713858.1 | 14 | 1877 | ||
GenBank mRNA | AF309777.1 | - | 15 | 1963 | |
AF367998.1 | - | 15 | 1963 | ||
AK092293.1 | - | 14 | 1569 | ||
AK315031.1 | - | 15 | 1902 | ||
BC049210.1 | - | 15 | 1927 | 5574223 | |
KU178816.1 | - | 15 | 1856 | ||
KU178817.1 | - | 14 | 1735 |