Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297732.1 | NP_001284661.1 | 3 | 592 | |
NM_001297733.1 | NP_001284662.1 | 3 | 596 | ||
NM_001300729.1 | NP_001287658.1 | 2 | 491 | ||
NM_173827.3 | NP_776188.1 | 3 | 592 | ||
XM_005265680.4 | XP_005265737.1 | 3 | 592 | ||
XM_011531878.2 | XP_011530180.1 | 3 | 563 | ||
XM_017008045.1 | XP_016863534.1 | 3 | 592 | ||
GenBank mRNA | AK096310.2 | - | 3 | 538 | |
AM055750.1 | - | 3 | 242 | ||
AM055751.1 | - | 3 | 246 | ||
AM055752.1 | - | 2 | 141 | ||
AY957564.1 | - | 3 | 588 | ||
AY957565.1 | - | 3 | 588 | ||
BC101684.1 | - | 3 | 525 | 8069190 | |
BC143642.1 | - | 3 | 525 | 9052156 |