Gene Transcripts

Gene Symbol
SHB
Entrez Gene ID
6461
Gene Name
SH2 domain containing adaptor protein B
Gene Aliases
bA3J10.2
Chromosome Location
Chr. 9: 37915898 - 38069213 on Build GRCh38
UniGene ID
Hs.521482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003028.2 NP_003019.2 3 1453
GenBank mRNA BC136581.1 - 3 1326
BC136582.1 - 3 1326 9020572
X75342.1 - 3 1456

View Full Product Details