Gene Transcripts

Gene Symbol
MND1
Entrez Gene ID
84057
Gene Name
meiotic nuclear divisions 1
Gene Aliases
GAJ
Chromosome Location
Chr. 4: 153344649 - 153415097 on Build GRCh38
UniGene ID
Hs.294088
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032117.3 NP_115493.1 6 475
NR_045605.1 8 636
XM_005263275.2 XP_005263332.1 6 434
XM_017008688.1 XP_016864177.1 6 832
GenBank mRNA AK312172.1 - 6 434
AY028916.1 - 6 414
BC032142.2 - 6 429 5095005
BF975093.1 - 8 604
BG758047.1 - 8 605
BM820228.1 - 6 475
BU533354.1 - 3 131

View Full Product Details