Gene Transcripts

Gene Symbol
RNF38
Entrez Gene ID
152006
Gene Name
ring finger protein 38
Gene Aliases
Chromosome Location
Chr. 9: 36336398 - 36487384 on Build GRCh38
UniGene ID
Hs.333503
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022781.4 NP_073618.3 12 3542
NM_194328.2 NP_919309.1 12 3709
NM_194329.2 NP_919310.1 11 3392
NM_194330.2 NP_919311.1 12 3440
NM_194332.2 NP_919313.1 11 3559
XM_005251366.3 XP_005251423.1 12 3546
XM_005251367.3 XP_005251424.1 12 3444
XM_005251368.3 XP_005251425.1 12 3558
XM_006716721.3 XP_006716784.1 11 3406
XM_011517712.2 XP_011516014.1 13 3482
XM_011517713.2 XP_011516015.1 12 3546
XM_017014294.1 XP_016869783.1 11 3463
XM_017014295.1 XP_016869784.1 12 3573
XM_017014296.1 XP_016869785.1 11 3143
XM_017014297.1 XP_016869786.1 11 3139
GenBank mRNA AF394047.1 - 12 3664
AK024996.1 - 10 3052
AK025466.1 - 1 1569

View Full Product Details