Gene Transcripts

Gene Symbol
SNRNP48
Entrez Gene ID
154007
Gene Name
small nuclear ribonucleoprotein U11/U12 subunit 48
Gene Aliases
C6orf151, dJ336K20B.1, dJ512B11.2
Chromosome Location
Chr. 6: 7590199 - 7611967 on Build GRCh38
UniGene ID
Hs.13366
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152551.3 NP_689764.3 9 3016
XM_011514312.2 XP_011512614.1 9 2952
GenBank mRNA AJ420586.1 - 1 1146
AK027220.1 - 4 2359
AL832462.1 - 9 3005
BC001558.1 - 1 531
BC114630.1 - 9 2984 40084896
BC114631.1 - 9 2985 40084903

View Full Product Details