Gene Transcripts

Gene Symbol
MRPL54
Entrez Gene ID
116541
Gene Name
mitochondrial ribosomal protein L54
Gene Aliases
L54mt, MRP-L54
Chromosome Location
Chr. 19: 3762667 - 3767565 on Build GRCh38
UniGene ID
Hs.356578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172251.2 NP_758455.1 3 518
GenBank mRNA BC065273.1 - 3 511
BF346319.1 - 3 511
BM792364.1 - 3 478
BQ061807.1 - 3 506

View Full Product Details