Gene Transcripts

Gene Symbol
MAGEB6
Entrez Gene ID
158809
Gene Name
MAGE family member B6
Gene Aliases
CT3.4, MAGE-B6, MAGEB6A
Chromosome Location
Chr. X: 26192440 - 26195646 on Build GRCh38
UniGene ID
Hs.376011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173523.2 NP_775794.2 2 1351
GenBank mRNA AK097561.1 - 2 1351
BC067286.1 - 2 1351 30386334
BC074920.2 - 1 1239 30915342

View Full Product Details