Gene Transcripts

Gene Symbol
TRIM63
Entrez Gene ID
84676
Gene Name
tripartite motif containing 63
Gene Aliases
IRF, MURF1, MURF2, RNF28, SMRZ
Chromosome Location
Chr. 1: 26051304 - 26067634 on Build GRCh38
UniGene ID
Hs.279709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032588.3 NP_115977.2 9 1441
GenBank mRNA AF353673.1 - 9 1420
AJ276484.1 - 9 1424
AK056942.1 - 9 1437
BC080529.1 - 9 1419 4862321

View Full Product Details