Gene Transcripts

Gene Symbol
SLC35B4
Entrez Gene ID
84912
Gene Name
solute carrier family 35 member B4
Gene Aliases
YEA, YEA4
Chromosome Location
Chr. 7: 134289337 - 134320307 on Build GRCh38
UniGene ID
Hs.490181
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032826.4 NP_116215.1 2, 3 507
XM_011516645.2 XP_011514947.1 2, 3 275
GenBank mRNA AB052892.1 - 2, 3 483
AJ315497.1 - 2, 3 339
AJ315498.1 - 2, 3 329
AJ971941.1 - 2, 3 183
AK027603.1 - 2, 3 347
AK027726.1 - 1, 2 65
AK075368.1 - 2, 3 312
AK222874.1 - 2, 3 356
AK307621.1 - 2, 3 335
BC008413.1 - 2, 3 328 4333393
DA208815.1 - 2, 3 507

View Full Product Details