Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281746.1 | NP_001268675.1 | 11 | 2683 | |
NM_001281747.1 | NP_001268676.1 | 11 | 2665 | ||
NM_138569.2 | NP_612636.2 | 10 | 1111 | ||
XM_005249476.4 | XP_005249533.1 | 11 | 2636 | ||
XM_005249478.4 | XP_005249535.1 | 10 | 2490 | ||
XM_005249480.4 | XP_005249537.1 | 10 | 1064 | ||
XM_006715245.3 | XP_006715308.2 | 11 | 2775 | ||
XM_017011465.1 | XP_016866954.1 | 9 | 2628 | ||
XM_017011466.1 | XP_016866955.1 | 9 | 2625 | ||
XM_017011467.1 | XP_016866956.1 | 8 | 2568 | ||
XM_017011468.1 | XP_016866957.1 | 9 | 2633 | ||
XM_017011469.1 | XP_016866958.1 | 9 | 2425 | ||
XM_017011470.1 | XP_016866959.1 | 8 | 1151 | ||
GenBank mRNA | AK055530.1 | - | 10 | 1036 | |
AK294890.1 | - | 11 | 2636 | ||
AK311190.1 | - | 11 | 2636 | ||
BC009010.1 | - | 5 | 576 | 4158300 | |
BX113168.1 | - | 3 | 114 | ||
BX647567.1 | - | 11 | 2629 | ||
KU178822.1 | - | 5 | 530 | ||
KU178823.1 | - | 8 | 824 | ||
KU178824.1 | - | 6 | 686 | ||
KU178825.1 | - | 7 | 752 | ||
KU178826.1 | - | 6 | 686 |