Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144578.3 | NP_653179.1 | 4 | 1729 | |
XM_011537362.2 | XP_011535664.2 | 4 | 2573 | ||
GenBank mRNA | AK025580.1 | - | 4 | 1693 | |
AL834285.1 | - | 4 | 1712 | ||
BC015621.2 | - | 4 | 1690 | 4843370 | |
U90909.1 | - | 1 | 724 |