Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284358.1 | NP_001271287.1 | 5 | 753 | |
NM_001284361.1 | NP_001271290.1 | 7 | 936 | ||
NM_001284362.1 | NP_001271291.1 | 7 | 936 | ||
NM_033030.5 | NP_149019.1 | 7 | 849 | ||
NM_197970.2 | NP_932074.1 | 7 | 786 | ||
XM_006712715.3 | XP_006712778.1 | 7 | 1146 | ||
XM_011511692.2 | XP_011509994.1 | 7 | 1146 | ||
XM_011511693.2 | XP_011509995.1 | 7 | 1146 | ||
XM_011511694.2 | XP_011509996.1 | 7 | 753 | ||
XM_017004773.1 | XP_016860262.1 | 7 | 1146 | ||
GenBank mRNA | AF272858.1 | - | 7 | 794 | |
AK058099.1 | - | 7 | 849 | ||
AK093453.1 | - | 7 | 936 | ||
AK097795.1 | - | 7 | 1145 | ||
AK098824.1 | - | 7 | 818 | ||
AK302817.1 | - | 5 | 753 | ||
AM295005.1 | - | 7 | 936 | ||
BC033674.1 | - | 7 | 801 | 5170732 |