Gene Transcripts

Gene Symbol
TTC12
Entrez Gene ID
54970
Gene Name
tetratricopeptide repeat domain 12
Gene Aliases
TPARM
Chromosome Location
Chr. 11: 113314529 - 113385952 on Build GRCh38
UniGene ID
Hs.288772
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318533.1 NP_001305462.1 9, 10 768
NM_017868.3 NP_060338.3 9, 10 750
XM_005271604.3 XP_005271661.1 9, 10 966
XM_011542890.2 XP_011541192.1 10, 11 964
XM_017017943.1 XP_016873432.1 9, 10 750
XM_017017944.1 XP_016873433.1 10, 11 1320
XM_017017945.1 XP_016873434.1 10, 11 803
XM_017017946.1 XP_016873435.1 9, 10 973
XM_017017947.1 XP_016873436.1 9, 10 750
XM_017017948.1 XP_016873437.1 9, 10 750
XM_017017949.1 XP_016873438.1 9, 10 750
XM_017017950.1 XP_016873439.1 9, 10 750
GenBank mRNA AK000542.1 - 8, 9 642
AK023921.1 - 9, 10 696
AK091887.1 - 11, 12 1362
AK223117.1 - 8, 9 642
AK292331.1 - 9, 10 688
BC011546.1 - 8, 9 692
BC032355.1 - 8, 9 672 5219117

View Full Product Details