Gene Transcripts

Gene Symbol
TOMM22
Entrez Gene ID
56993
Gene Name
translocase of outer mitochondrial membrane 22
Gene Aliases
1C9-2, MST065, MSTP065, TOM22
Chromosome Location
Chr. 22: 38681949 - 38684760 on Build GRCh38
UniGene ID
Hs.595072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020243.4 NP_064628.1 4 720
GenBank mRNA AB041906.1 - 4 703
AK024731.1 - 4 712
BC009363.2 - 4 693 4302531
BQ961547.1 - 1 60

View Full Product Details