Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018518.4 | NP_060988.3 | 4 | 529 | |
NM_182751.2 | NP_877428.1 | 4 | 529 | ||
XM_011519538.2 | XP_011517840.1 | 4 | 499 | ||
GenBank mRNA | AB042719.1 | - | 4 | 496 | |
AK055695.1 | - | 4 | 502 | ||
AL136840.1 | - | 3 | 495 | ||
BC101727.1 | - | 4 | 498 | 8069233 | |
BC143490.1 | - | 4 | 494 | 9051998 | |
DA024424.1 | - | 4 | 529 |