Gene Transcripts

Gene Symbol
PLD3
Entrez Gene ID
23646
Gene Name
phospholipase D family member 3
Gene Aliases
AD19, HU-K4, HUK4
Chromosome Location
Chr. 19: 40348395 - 40378490 on Build GRCh38
UniGene ID
Hs.257008
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031696.3 NP_001026866.1 11 1567
NM_001291311.1 NP_001278240.1 11 1501
NM_012268.3 NP_036400.2 11 1791
XM_005258704.1 XP_005258761.1 11 1712
XM_005258707.4 XP_005258764.1 11 1494
XM_005258708.3 XP_005258765.1 10 1387
XM_005258709.4 XP_005258766.1 10 1375
XM_005258710.4 XP_005258767.1 10 1416
XM_006723122.1 XP_006723185.1 11 1879
XM_011526692.1 XP_011524994.1 11 1874
XM_011526693.1 XP_011524995.1 11 1594
XM_017026546.1 XP_016882035.1 12 1810
XM_017026547.1 XP_016882036.1 11 1378
XM_017026548.1 XP_016882037.1 11 1529
XM_017026549.1 XP_016882038.1 11 1632
GenBank mRNA AK293690.1 - 11 1235
BC000553.1 - 7 1037
BC036327.1 - 11 1292 5189261
BC096820.1 - 11 1490 30558682
U60644.1 - 11 1497

View Full Product Details