Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001191322.1 | NP_001178251.1 | 3 | 1201 | |
NM_001191323.1 | NP_001178252.1 | 3 | 1288 | ||
NM_013372.6 | NP_037504.1 | 2 | 1411 | ||
XM_005254301.2 | XP_005254358.1 | 2 | 1475 | ||
XM_017022077.1 | XP_016877566.1 | 2 | 1457 | ||
GenBank mRNA | AF110137.2 | - | 2 | 1314 | |
AF154054.1 | - | 2 | 1380 | ||
AK095741.1 | - | 2 | 1255 | ||
AK095890.1 | - | 3 | 1288 | ||
AK095972.1 | - | 3 | 1201 | ||
AY232290.1 | - | 2 | 1387 |