Gene Transcripts
- Gene Symbol
- PRICKLE4
- Entrez Gene ID
- 29964
- Gene Name
- prickle planar cell polarity protein 4
- Gene Aliases
- C6orf49, OBTP, OEBT, TOMM6
- Chromosome Location
- Chr. 6: 41780762 - 41787372 on Build GRCh38
- UniGene ID
- Hs.715653
- Species
- Homo sapiens
- Gene Symbol
- TOMM6
- Entrez Gene ID
- 100188893
- Gene Name
- translocase of outer mitochondrial membrane 6
- Gene Aliases
- OBTP, TOM6
- Chromosome Location
- Chr. 6: 41787443 - 41789896 on Build GRCh38
- UniGene ID
- Hs.731633
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AJ420506.1 |
- |
1 |
303 |
|
View Full Product Details