Gene Transcripts

Gene Symbol
OSTM1
Entrez Gene ID
28962
Gene Name
osteopetrosis associated transmembrane protein 1
Gene Aliases
GIPN, GL, HSPC019, OPTB5
Chromosome Location
Chr. 6: 108041409 - 108074737 on Build GRCh38
UniGene ID
Hs.226780
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014028.3 NP_054747.2 6 2513
GenBank mRNA AF077205.1 - 4 1866
AF533891.1 - 6 2497
AJ420489.1 - 1 884
AJ420572.1 - 1 1177
AK075012.1 - 6 2439
AK129854.1 - 6 2507
BC068581.1 - 6 2495 5298450

View Full Product Details