Gene Transcripts
- Gene Symbol
- STK36
- Entrez Gene ID
- 27148
- Gene Name
- serine/threonine kinase 36
- Gene Aliases
- FU
- Chromosome Location
- Chr. 2: 218672026 - 218702717 on Build GRCh38
- UniGene ID
- Hs.471404
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243313.1 |
NP_001230242.1
|
17, 18 |
2426 |
|
NM_015690.4 |
NP_056505.2
|
17, 18 |
2426 |
|
XM_005246464.1 |
XP_005246521.1
|
17, 18 |
2314 |
|
XM_011510959.1 |
XP_011509261.1
|
17, 18 |
2399 |
|
XM_011510960.1 |
XP_011509262.1
|
17, 18 |
2367 |
|
XM_011510961.1 |
XP_011509263.1
|
8, 9 |
997 |
|
XM_017003804.1 |
XP_016859293.1
|
17, 18 |
2314 |
|
XM_017003805.1 |
XP_016859294.1
|
16, 17 |
2103 |
|
GenBank mRNA |
AB033104.1 |
- |
17, 18 |
2383 |
|
AF200815.1 |
- |
16, 17 |
2147 |
|
BC026158.2 |
- |
17, 18 |
2366 |
4839214 |
View Full Product Details