Gene Transcripts

Gene Symbol
MRPS33
Entrez Gene ID
51650
Gene Name
mitochondrial ribosomal protein S33
Gene Aliases
CGI-139, MRP-S33, PTD003, S33mt
Chromosome Location
Chr. 7: 141006161 - 141014981 on Build GRCh38
UniGene ID
Hs.416207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016071.3 NP_057155.1 2 329
NM_053035.2 NP_444263.1 2 271
GenBank mRNA AF078858.1 - 2 245
AF151897.1 - 2 307
AK024776.1 - 2 327
BC015462.2 - 1 202 3889203
BC105793.1 - 2 240
BQ420818.1 - 2 271
W95986.1 - 1 212

View Full Product Details