Gene Transcripts

Gene Symbol
ANKRD17
Entrez Gene ID
26057
Gene Name
ankyrin repeat domain 17
Gene Aliases
GTAR, MASK2, NY-BR-16
Chromosome Location
Chr. 4: 73073966 - 73259142 on Build GRCh38
UniGene ID
Hs.601206
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286771.2 NP_001273700.1 34 8383
NM_015574.1 NP_056389.1 34 8762
NM_032217.4 NP_115593.3 34 8765
NM_198889.2 NP_942592.1 33 8012
XM_005265671.3 XP_005265728.1 33 8353
XM_017008011.1 XP_016863500.1 34 8337
XM_017008012.1 XP_016863501.1 33 7587
XM_017008013.1 XP_016863502.1 33 7584
GenBank mRNA AB014597.2 - 34 8286

View Full Product Details