Gene Transcripts
- Gene Symbol
- RASGRP3
- Entrez Gene ID
- 25780
- Gene Name
- RAS guanyl releasing protein 3
- Gene Aliases
- GRP3
- Chromosome Location
- Chr. 2: 33436348 - 33564731 on Build GRCh38
- UniGene ID
- Hs.143674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001139488.1 |
NP_001132960.1
|
13, 14 |
2117 |
|
NM_015376.2 |
NP_056191.1
|
12, 13 |
1672 |
|
NM_170672.2 |
NP_733772.1
|
14, 15 |
2030 |
|
XM_005264246.3 |
XP_005264303.1
|
13, 14 |
2454 |
|
XM_011532746.2 |
XP_011531048.1
|
14, 15 |
1904 |
|
XM_011532747.2 |
XP_011531049.1
|
13, 14 |
1919 |
|
XM_011532748.2 |
XP_011531050.2
|
13, 14 |
1765 |
|
XM_017003759.1 |
XP_016859248.1
|
15, 16 |
3380 |
|
XM_017003760.1 |
XP_016859249.1
|
14, 15 |
3852 |
|
XM_017003761.1 |
XP_016859250.1
|
13, 14 |
1782 |
|
XM_017003762.1 |
XP_016859251.1
|
14, 15 |
2028 |
|
XM_017003763.1 |
XP_016859252.1
|
13, 14 |
1758 |
|
GenBank mRNA |
AB020653.1 |
- |
12, 13 |
1672 |
|
AK292983.1 |
- |
14, 15 |
1941 |
|
BC027849.1 |
- |
14, 15 |
2030 |
5203041 |
BX647990.1 |
- |
13, 14 |
1725 |
|
View Full Product Details