Gene Transcripts

Gene Symbol
MAB21L2
Entrez Gene ID
10586
Gene Name
mab-21 like 2
Gene Aliases
MCOPS14
Chromosome Location
Chr. 4: 150581925 - 150584693 on Build GRCh38
UniGene ID
Hs.584852
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006439.4 NP_006430.1 1 2300
GenBank mRNA AF262032.1 - 1 1950
AK055665.1 - 1 2074
BC009983.2 - 1 1798 4125321

View Full Product Details