Gene Transcripts
- Gene Symbol
- MTX2
- Entrez Gene ID
- 10651
- Gene Name
- metaxin 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 176269395 - 176338025 on Build GRCh38
- UniGene ID
- Hs.470728
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001006635.2 |
NP_001006636.1
|
10, 11 |
1065 |
|
NM_001319097.1 |
NP_001306026.1
|
9, 10 |
792 |
|
NM_006554.4 |
NP_006545.1
|
9, 10 |
861 |
|
XM_011510497.2 |
XP_011508799.1
|
10, 11 |
958 |
|
GenBank mRNA |
AF053551.1 |
- |
9, 10 |
834 |
|
AK303564.1 |
- |
10, 11 |
990 |
|
AK315918.1 |
- |
10, 11 |
1001 |
|
BC017271.2 |
- |
10, 11 |
995 |
4635884 |
BC067831.1 |
- |
9, 10 |
840 |
5298149 |
BC088359.1 |
- |
9, 10 |
796 |
30378040 |
CX165048.1 |
- |
9, 10 |
685 |
|
View Full Product Details