Gene Transcripts

Gene Symbol
WDHD1
Entrez Gene ID
11169
Gene Name
WD repeat and HMG-box DNA binding protein 1
Gene Aliases
AND-1, AND1, CHTF4, CTF4
Chromosome Location
Chr. 14: 54938938 - 55027105 on Build GRCh38
UniGene ID
Hs.385998
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008396.2 NP_001008397.1 9 822
NM_007086.3 NP_009017.1 10 915
XM_006720012.2 XP_006720075.1 10 913
XM_011536373.2 XP_011534675.1 9 826
XM_017020940.1 XP_016876429.1 9 826
XM_017020941.1 XP_016876430.1 7 636
GenBank mRNA AJ006266.1 - 9 864
AK293005.1 - 10 882
BC000622.2 - 10 880
BC005040.1 - 10 880
BC043349.1 - 10 903
BC063041.1 - 10 891 6514641

View Full Product Details