Gene Transcripts

Gene Symbol
CIT
Entrez Gene ID
11113
Gene Name
citron rho-interacting serine/threonine kinase
Gene Aliases
CRIK, STK21
Chromosome Location
Chr. 12: 119685790 - 119877318 on Build GRCh38
UniGene ID
Hs.119594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206999.1 NP_001193928.1 5 499
NM_007174.2 NP_009105.1 5 499
XM_006719206.2 XP_006719269.1 5 526
XM_011537783.1 XP_011536085.1 5 526
XM_011537784.1 XP_011536086.1 5 526
XM_011537785.1 XP_011536087.1 5 526
XM_011537787.1 XP_011536089.1 5 526
XM_011537788.1 XP_011536090.1 5 526
XM_017018735.1 XP_016874224.1 5 526
XM_017018736.1 XP_016874225.1 5 526
XM_017018737.1 XP_016874226.1 5 526
GenBank mRNA AY209000.1 - 5 496
AY257469.1 - 5 496
AY681966.1 - 5 499

View Full Product Details