Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr. 7: 128954180 - 129055173 on Build GRCh38
UniGene ID
Hs.193613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191028.2 NP_001177957.2 4 898
NM_012470.3 NP_036602.1 4 898
NR_034053.2 5 1059
XM_011515989.2 XP_011514291.1 4 500
GenBank mRNA AF145029.1 - 4 608
AJ133769.1 - 4 495
AK225999.1 - 4 869
AK312580.1 - 4 625
BC068530.1 - 4 626

View Full Product Details