Gene Transcripts

Gene Symbol
CDC16
Entrez Gene ID
8881
Gene Name
cell division cycle 16
Gene Aliases
ANAPC6, APC6, CDC16Hs, CUT9
Chromosome Location
Chr. 13: 114234845 - 114272675 on Build GRCh38
UniGene ID
Hs.374127
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001078645.1 NP_001072113.1 5 562
NM_001318517.1 NP_001305446.1 5 559
NM_001318518.1 NP_001305447.1 5 559
NM_003903.3 NP_003894.3 5 562
XM_005266206.1 XP_005266263.1 5 604
XM_006719992.1 XP_006720055.1 5 549
XM_006719993.1 XP_006720056.1 4 497
XM_011534851.1 XP_011533153.1 5 604
XM_011534853.1 XP_011533155.1 5 604
XM_017020830.1 XP_016876319.1 5 604
XM_017020831.1 XP_016876320.1 5 604
XM_017020832.1 XP_016876321.1 4 497
XM_017020833.1 XP_016876322.1 3 182
GenBank mRNA AB209850.1 - 4 1715
AF164598.1 - 5 365
AK296424.1 - 5 552
AK315302.1 - 5 393
AL540490.3 - 5 559
BC010875.2 - 5 506 3879150
BC017244.1 - 5 472 4877392
U18291.1 - 5 385

View Full Product Details