Gene Transcripts

Gene Symbol
SCAMP1
Entrez Gene ID
9522
Gene Name
secretory carrier membrane protein 1
Gene Aliases
SCAMP, SCAMP37
Chromosome Location
Chr. 5: 78360503 - 78480739 on Build GRCh38
UniGene ID
Hs.482587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290229.1 NP_001277158.1 8 2062
NM_004866.5 NP_004857.4 9 2140
NR_110885.1 8 1980
XM_011543727.2 XP_011542029.1 8 2038
GenBank mRNA AK001541.1 - 3 1368
AK225172.1 - 9 2029
BC016355.1 - 3 1398
BC034048.1 - 9 1994 4814951

View Full Product Details