Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004890.2 | NP_004881.2 | 7 | 864 | |
XM_006721600.2 | XP_006721663.1 | 7 | 1489 | ||
GenBank mRNA | AF047437.1 | - | 7 | 840 | |
AK123065.1 | - | 1643 | |||
AK312174.1 | - | 7 | 840 | ||
BC011934.2 | - | 7 | 833 | 4330461 |