Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005443.4 | NP_005434.4 | 12 | 1989 | |
XM_011532400.1 | XP_011530702.1 | 12 | 1969 | ||
XM_011532401.1 | XP_011530703.1 | 12 | 2055 | ||
GenBank mRNA | AF033026.1 | - | 12 | 1931 | |
AF105227.1 | - | 12 | 1959 | ||
AK292774.1 | - | 12 | 1939 | ||
BC011392.1 | - | 12 | 1862 | 3869484 | |
BC040940.1 | - | 12 | 1917 | ||
BC050627.1 | - | 12 | 1925 | 5556165 | |
BU517185.1 | - | 2 | 253 | ||
Y10387.1 | - | 12 | 1941 |