Gene Transcripts

Gene Symbol
XAGE1B
Entrez Gene ID
653220
Gene Name
X antigen family member 1B
Gene Aliases
CT12.1, CT12.1A, CT12.1B, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1A
Chromosome Location
Chr. X: 52495668 - 52500815 on Build GRCh38
UniGene ID
Hs.112208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097594.2 NP_001091063.2 3 421
NM_001097596.2 NP_001091065.1 3 284
NR_033254.1 3 532
XM_017029748.1 XP_016885237.1 3 553
XM_017029749.1 XP_016885238.1 3 421

Gene Symbol
XAGE1E
Entrez Gene ID
653067
Gene Name
X antigen family member 1E
Gene Aliases
CT12.1, CT12.1C, CT12.1D, CT12.1E, CTP9, GAGED2, XAGE-1, XAGE1, XAGE1C, XAGE1D
Chromosome Location
Chr. X: 52512074 - 52517221 on Build GRCh38
UniGene ID
Hs.112208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001097604.2 NP_001091073.2 3 421
NM_001097605.2 NP_001091074.1 3 284
NR_033257.1 3 532
XM_017029746.1 XP_016885235.1 3 553
XM_017029747.1 XP_016885236.1 3 421
GenBank mRNA AF251237.1 - 3 413
AJ290447.1 - 3 257
AJ318878.1 - 3 532
AJ318879.1 - 3 273
BC009538.1 - 3 316
BT007099.1 - 2 133

View Full Product Details