Gene Transcripts

Gene Symbol
TARBP2
Entrez Gene ID
6895
Gene Name
TARBP2, RISC loading complex RNA binding subunit
Gene Aliases
LOQS, TRBP, TRBP1, TRBP2
Chromosome Location
Chr. 12: 53500921 - 53506431 on Build GRCh38
UniGene ID
Hs.326
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004178.4 NP_004169.3 2 200
NM_134323.1 NP_599150.1 2 665
NM_134324.2 NP_599151.2 2 240
XM_005269114.1 XP_005269171.1 2 235
XM_005269115.2 XP_005269172.1 2 242
XM_005269117.1 XP_005269174.1 2 284
XM_005269120.4 XP_005269177.1 2 665
XM_005269122.3 XP_005269179.1 2 237
XM_011538712.2 XP_011537014.1 2 223
XM_017019910.1 XP_016875399.1 2 235
XM_017019911.1 XP_016875400.1 2 295
GenBank mRNA AK290737.1 - 2 238
AK293728.1 - 2 236
AK309569.1 - 2 284
BC002419.2 - 2 665
BC005860.2 - 2 530 2960701
BI771121.1 - 2 243
BT007140.1 - 2 177
M60801.1 - 2 168
U08998.2 - 2 265

View Full Product Details