Gene Transcripts
- Gene Symbol
- IL32
- Entrez Gene ID
- 9235
- Gene Name
- interleukin 32
- Gene Aliases
- IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
- Chromosome Location
- Chr. 16: 3065312 - 3069819 on Build GRCh38
- UniGene ID
- Hs.943
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001012631.1 |
NP_001012649.1
|
7, 8 |
490 |
|
NM_001012632.1 |
NP_001012650.1
|
6, 7 |
289 |
|
NM_001012633.1 |
NP_001012651.1
|
6, 7 |
423 |
|
NM_001012634.1 |
NP_001012652.1
|
5, 6 |
230 |
|
NM_001012635.1 |
NP_001012653.1
|
5, 6 |
225 |
|
NM_001012636.1 |
NP_001012654.1
|
4, 5 |
213 |
|
NM_001012718.1 |
NP_001012736.1
|
6, 7 |
426 |
|
NM_001308078.1 |
NP_001295007.1
|
5, 6 |
424 |
|
NM_004221.4 |
NP_004212.4
|
6, 7 |
423 |
|
GenBank mRNA |
AK296143.1 |
- |
5, 6 |
235 |
|
AY495331.1 |
- |
6, 7 |
413 |
|
AY495332.1 |
- |
6, 7 |
413 |
|
AY495333.1 |
- |
5, 6 |
371 |
|
BC009401.2 |
- |
5, 6 |
561 |
4300407 |
BC018782.1 |
- |
6, 7 |
270 |
4873608 |
BC105602.1 |
- |
6, 7 |
291 |
6496101 |
BF568456.1 |
- |
5, 6 |
562 |
|
BG748458.1 |
- |
6, 7 |
369 |
|
BM759359.1 |
- |
3, 4 |
164 |
|
BM822755.1 |
- |
7, 8 |
398 |
|
EU339107.1 |
- |
3, 4 |
92 |
|
FJ985780.1 |
- |
5, 6 |
317 |
|
JN546102.1 |
- |
4, 5 |
152 |
|
M59807.1 |
- |
5, 6 |
409 |
|
View Full Product Details