Gene Transcripts

Gene Symbol
DYM
Entrez Gene ID
54808
Gene Name
dymeclin
Gene Aliases
DMC, SMC
Chromosome Location
Chr. 18: 49041474 - 49460709 on Build GRCh38
UniGene ID
Hs.162996
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017653.3 NP_060123.3 8, 9 1127
XM_006722488.2 XP_006722551.1 8, 9 1122
XM_006722490.2 XP_006722553.1 8, 9 1122
XM_006722491.2 XP_006722554.1 8, 9 1122
XM_006722492.3 XP_006722555.1 8, 9 1122
XM_011526036.1 XP_011524338.1 8, 9 1122
XM_011526037.1 XP_011524339.1 8, 9 1124
XM_011526039.1 XP_011524341.1 8, 9 1122
XM_011526040.1 XP_011524342.1 9, 10 1123
XM_011526041.1 XP_011524343.1 8, 9 1122
XM_011526042.1 XP_011524344.1 8, 9 1122
XM_017025796.1 XP_016881285.1 9, 10 1133
XM_017025797.1 XP_016881286.1 8, 9 1060
XM_017025799.1 XP_016881288.1 8, 9 1122
GenBank mRNA AK000078.1 - 4, 5 440
AK074611.1 - 8, 9 1122
AK091256.1 - 9, 10 1096
AK225205.1 - 4, 5 440
AK291303.1 - 8, 9 1092
AK315091.1 - 9, 10 1068
AY364250.1 - 7, 8 762
BC001252.1 - 5, 6 526
BC064394.1 - 8, 9 1109 5752466

View Full Product Details