Gene Transcripts

Gene Symbol
THUMPD1
Entrez Gene ID
55623
Gene Name
THUMP domain containing 1
Gene Aliases
Tan1
Chromosome Location
Chr. 16: 20730255 - 20741964 on Build GRCh38
UniGene ID
Hs.700004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017736.4 NP_060206.2 4 1473
XM_011545891.1 XP_011544193.1 5 1257
GenBank mRNA AK000281.1 - 4 1327
BC000448.2 - 4 1305 2821354

View Full Product Details