Gene Transcripts

Gene Symbol
NUFIP2
Entrez Gene ID
57532
Gene Name
NUFIP2, FMR1 interacting protein 2
Gene Aliases
182-FIP, 82-FIP, FIP-82, PIG1
Chromosome Location
Chr. 17: 29255836 - 29294148 on Build GRCh38
UniGene ID
Hs.462598
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020772.2 NP_065823.1 2 481
XM_017024896.1 XP_016880385.1 2 1779
GenBank mRNA AB037742.1 - 2 451
AJ493465.1 - 2 392
AK293075.1 - 2 481
AY232289.1 - 2 460
BC108307.1 - 2 438 6502841
BC129990.1 - 2 396 40128910

View Full Product Details