Gene Transcripts

Gene Symbol
APOPT1
Entrez Gene ID
84334
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
APOP, APOP1, C14orf153
Chromosome Location
Chr. 14: 103562957 - 103592187 on Build GRCh38
UniGene ID
Hs.598441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302652.1 NP_001289581.1 5 841
NM_001302653.1 NP_001289582.1 6 931
NM_032374.4 NP_115750.2 5 843
NR_126431.1 6 1008
NR_126432.1 4 779
GenBank mRNA AK223092.1 - 5 814
AK223103.1 - 5 814
BC007412.2 - 5 806 2989951
BP204024.1 - 3 567
BU956674.1 - 4 736

View Full Product Details