Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166261.1 | NP_001159733.1 | 4 | 695 | |
NM_001166262.1 | NP_001159734.1 | 4 | 765 | ||
NM_001166263.1 | NP_001159735.1 | 5 | 794 | ||
NM_152912.4 | NP_690876.3 | 4 | 733 | ||
XM_006719771.3 | XP_006719834.1 | 5 | 781 | ||
XM_006719772.3 | XP_006719835.1 | 4 | 1012 | ||
XM_011534957.2 | XP_011533259.1 | 5 | 1255 | ||
XM_011534958.2 | XP_011533260.1 | 5 | 1039 | ||
XM_011534959.2 | XP_011533261.1 | 5 | 1204 | ||
XM_011534960.2 | XP_011533262.1 | 5 | 869 | ||
XM_011534961.2 | XP_011533263.1 | 5 | 1457 | ||
XM_011534962.2 | XP_011533264.1 | 5 | 1034 | ||
XM_011534963.2 | XP_011533265.1 | 5 | 1622 | ||
XM_017020417.1 | XP_016875906.1 | 4 | 639 | ||
GenBank mRNA | AF410851.1 | - | 3 | 600 | |
AK094420.1 | - | 2 | 1794 | ||
BC039599.1 | - | 4 | 686 | 3855096 | |
BC046166.1 | - | 4 | 705 | 3460180 | |
BM548061.1 | - | 3 | 658 | ||
BM768315.1 | - | 2 | 179 |