Gene Transcripts
- Gene Symbol
- POC1B
- Entrez Gene ID
- 282809
- Gene Name
- POC1 centriolar protein B
- Gene Aliases
- CORD20, PIX1, TUWD12, WDR51B
- Chromosome Location
- Chr. 12: 89419718 - 89526262 on Build GRCh38
- UniGene ID
- Hs.25130
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199777.1 |
NP_001186706.1
|
5, 6 |
1022 |
|
NM_172240.2 |
NP_758440.1
|
6, 7 |
1041 |
|
NR_037659.1 |
|
5, 6 |
869 |
|
NR_037660.1 |
|
4, 5 |
850 |
|
GenBank mRNA |
AB053266.1 |
- |
5, 6 |
531 |
|
AK027829.1 |
- |
2, 3 |
240 |
|
AK074772.1 |
- |
6, 7 |
779 |
|
AK123106.1 |
- |
5, 6 |
1019 |
|
AK295596.1 |
- |
5, 6 |
607 |
|
AL832918.1 |
- |
4, 5 |
506 |
|
BC026080.2 |
- |
6, 7 |
803 |
4827689 |
BU521431.1 |
- |
4, 5 |
519 |
|
EF011619.1 |
- |
6, 7 |
803 |
|
View Full Product Details