Gene Transcripts

Gene Symbol
SGCA
Entrez Gene ID
6442
Gene Name
sarcoglycan alpha
Gene Aliases
50DAG, ADL, DAG2, DMDA2, LGMD2D, SCARMD1, adhalin
Chromosome Location
Chr. 17: 50165517 - 50175932 on Build GRCh38
UniGene ID
Hs.463412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000023.3 NP_000014.1 10 1338
NM_001135697.2 NP_001129169.1 8 966
NR_135553.1 9 1129
XM_011525120.2 XP_011523422.2 10 1730
XM_011525121.2 XP_011523423.2 10 1582
XM_011525122.2 XP_011523424.2 9 1521
XM_011525123.2 XP_011523425.2 8 1358
XM_011525124.2 XP_011523426.1 9 1085
GenBank mRNA AK290622.1 - 10 1311
BC025702.1 - 10 1310 5222483
F25770.1 - 2 280
L35853.1 - 8 947
U08895.1 - 10 1293

View Full Product Details