Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 778 | |
XM_006716463.2 | XP_006716526.1 | 6, 7 | 1371 | ||
GenBank mRNA | AK225246.1 | - | 1 | 777 | |
AK225254.1 | - | 1 | 776 | ||
AK291119.1 | - | 6, 7 | 776 | ||
BC006095.1 | - | 1 | 766 | 2821364 | |
BC008850.2 | - | 1 | 803 | 4138113 | |
BC009599.1 | - | 1 | 778 | 4134557 | |
BC071671.1 | - | 1 | 759 | 4392472 | |
BC071894.1 | - | 1 | 763 | 2906310 | |
BC071895.1 | - | 1 | 761 | 6568654 | |
BC087837.1 | - | 1 | 762 | 6170371 | |
L16558.1 | - | 1 | 756 | ||
X52967.1 | - | 6, 7 | 777 | ||
X57958.1 | - | 1 | 793 | ||
X57959.1 | - | 1 | 766 |